Learn More
Medchemexpress LLC (±)-Naringenin (Standard) | 67604-48-2 | MFCD00006844 | 98.8% | 272.25 | 50 MG
SDP
Catalog No. 5000455004
Encompass_Preferred
(±)-Naringenin (Standard) is an analytical standard of (±)-Naringenin, designed for research and analytical applications. This naturally-occurring flavonoid exhibits a vasorelaxant effect on endothelium-denuded vessels by activating BKCa channels in myocytes. As an analytical standard grade compound, it serves as a reliable reference for various assays.
  • Serves as a reference standard for assays.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-004-55004 Supplier Medchemexpress LLC Supplier No. HYW011641R50MG
(±)-Naringenin (Standard) is an analytical standard of (±)-Naringenin, designed for research and analytical applications. This naturally-occurring flavonoid exhibits a vasorelaxant effect on endothelium-denuded vessels by activating BKCa channels in myocytes. As an analytical standard grade compound, it serves as a reliable reference for various assays.
  • Serves as a reference standard for assays.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.