Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naringenin triacetate | 3682-04-0 | MFCD00706050 | >98.0% | 398.36 | C21H18O8 | 10 MG
SDP

Catalog No. 5000289381
Encompass_Preferred

Naringenin triacetate is a plant-derived flavonoid research reagent reported to bind the first bromodomain of BRD4 (BRD4 BD1). It is provided as a solid for laboratory in-vitro studies and is for research use only, not for human or animal administration.

  • Reported binder of BRD4 first bromodomain (BRD4 BD1).
  • CAS number 3682-04-0.
  • Molecular formula C21H18O8; molecular weight 398.36.
  • Available in small research quantities, e.g., 10 mg.
  • Purity reported >98% by HPLC from distributor sources.
  • Intended for in-vitro research use only.

Catalog No. 50-002-89381 Supplier Medchemexpress LLC Supplier No. HYN321310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naringenin triacetate is a plant-derived flavonoid research reagent reported to bind the first bromodomain of BRD4 (BRD4 BD1). It is provided as a solid for laboratory in-vitro studies and is for research use only, not for human or animal administration.

  • Reported binder of BRD4 first bromodomain (BRD4 BD1).
  • CAS number 3682-04-0.
  • Molecular formula C21H18O8; molecular weight 398.36.
  • Available in small research quantities, e.g., 10 mg.
  • Purity reported >98% by HPLC from distributor sources.
  • Intended for in-vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.