Learn More
Apexbio Technology LLC Naringin 10236-47-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094459
Encompass_Preferred
Naringin (CAS 10236-47-2) is a flavanone glycoside predominantly isolated from the fruit of Citrus paradisi Macfadyen (grapefruit) It is recognized for modulating various cellular pathways including inhibition of oxidative stress and inflammation through interaction with NF- B and MAPK signaling cascades Naringin has demonstrated antioxidative anti-inflammatory and potential anti-carcinogenic properties in cellular and animal models Widely used in biomedical research it serves as a tool compound for investigating mechanisms underlying chronic diseases metabolic syndromes and the modulation of drug-metabolizing enzymes For optimal stability it should be stored in a tightly sealed container under cool dry conditions
Catalog No. 50-000-94459 Supplier Apexbio Technology LLC Supplier No. N23715.1
Naringin (CAS 10236-47-2) is a flavanone glycoside predominantly isolated from the fruit of Citrus paradisi Macfadyen (grapefruit) It is recognized for modulating various cellular pathways including inhibition of oxidative stress and inflammation through interaction with NF- B and MAPK signaling cascades Naringin has demonstrated antioxidative anti-inflammatory and potential anti-carcinogenic properties in cellular and animal models Widely used in biomedical research it serves as a tool compound for investigating mechanisms underlying chronic diseases metabolic syndromes and the modulation of drug-metabolizing enzymes For optimal stability it should be stored in a tightly sealed container under cool dry conditions