Learn More
Apexbio Technology LLC Naringin 10236-47-2 200mg
SDP
Catalog No. 502240510
Encompass_Preferred
Naringin (CAS 10236-47-2) is a flavanone glycoside predominantly isolated from the fruit of Citrus paradisi Macfadyen (grapefruit) It is recognized for modulating various cellular pathways including inhibition of oxidative stress and inflammation through interaction with NF- B and MAPK signaling cascades Naringin has demonstrated antioxidative anti-inflammatory and potential anti-carcinogenic properties in cellular and animal models Widely used in biomedical research it serves as a tool compound for investigating mechanisms underlying chronic diseases metabolic syndromes and the modulation of drug-metabolizing enzymes For optimal stability it should be stored in a tightly sealed container under cool dry conditions
This item is not returnable. View return policy
Catalog No. 50-224-0510 Supplier Apexbio Technology LLC Supplier No. N2371200
Naringin (CAS 10236-47-2) is a flavanone glycoside predominantly isolated from the fruit of Citrus paradisi Macfadyen (grapefruit) It is recognized for modulating various cellular pathways including inhibition of oxidative stress and inflammation through interaction with NF- B and MAPK signaling cascades Naringin has demonstrated antioxidative anti-inflammatory and potential anti-carcinogenic properties in cellular and animal models Widely used in biomedical research it serves as a tool compound for investigating mechanisms underlying chronic diseases metabolic syndromes and the modulation of drug-metabolizing enzymes For optimal stability it should be stored in a tightly sealed container under cool dry conditions