Learn More
Medchemexpress LLC HY-N0153 200mg Medchemexpress, Naringin CAS:10236-47-2 Purity:>98%
SDP
Catalog No. 501933430
Encompass_Preferred
Medchemexpress, HY-N0153 200mg Naringin CAS:10236-47-2 Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3430 Supplier Medchemexpress LLC Supplier No. HYN0153200MG
Medchemexpress, HY-N0153 200mg Naringin CAS:10236-47-2 Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.