Learn More
Selleck Chemical LLC Naringin S2329-1g
SDP
Catalog No. 503112765
Encompass_Preferred
Naringin (Naringoside) is a flavanone glycoside which exerts a variety of pharmacological effects such as antioxidant activity blood lipid lowering anticancer activity and inhibition of cytochrome P450 enzymes
Catalog No. 50-311-2765 Supplier Selleck Chemical LLC Supplier No. S23291G
Naringin (Naringoside) is a flavanone glycoside which exerts a variety of pharmacological effects such as antioxidant activity blood lipid lowering anticancer activity and inhibition of cytochrome P450 enzymes