Learn More
Apexbio Technology LLC NARINGIN-DIHYDROCHALCONE-10G
SDP
Catalog No. 5000570450
Encompass_Preferred
Naringin Dihydrochalcone (CAS No 18916-17-1) is a flavonoid derivative commonly researched in the fields of pharmacology and nutrition It originates from citrus fruits particularly grapefruit and acts as a potent sweetening agent Its mechanism involves modulation of taste receptors and potential effects on cellular pathways related to oxidative stress In vitro it exhibits activity in the nanomolar to low micromolar range It is utilized in various experimental settings including cell models for antioxidant and anti-inflammatory studies as well as animal studies evaluating metabolic effects Drug screening often explores its potential in improving oral bioavailability and reducing bitterness in formulations Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70450 Supplier Apexbio Technology LLC Supplier No. C644710000
Naringin Dihydrochalcone (CAS No 18916-17-1) is a flavonoid derivative commonly researched in the fields of pharmacology and nutrition It originates from citrus fruits particularly grapefruit and acts as a potent sweetening agent Its mechanism involves modulation of taste receptors and potential effects on cellular pathways related to oxidative stress In vitro it exhibits activity in the nanomolar to low micromolar range It is utilized in various experimental settings including cell models for antioxidant and anti-inflammatory studies as well as animal studies evaluating metabolic effects Drug screening often explores its potential in improving oral bioavailability and reducing bitterness in formulations Experimental concentrations typically depend on the specific research design