Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naringin dihydrochalcone, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6683003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6683003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6683003 Supplier Thermo Scientific Chemicals Supplier No. J6683003
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18916-17-1 | Molecular Formula: C27H34O14 | Molecular Weight: 582.555 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18916-17-1
Molecular Formula C27H34O14
Molecular Weight (g/mol) 582.555
MDL Number MFCD08436145
InChI Key CWBZAESOUBENAP-NDAHTVSZSA-N
Synonym 1-[4-[(2-O-(6-Deoxy-a-L-rhamnopyranosyl)-b-D-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone
PubChem CID 126963586
IUPAC Name 1-[4-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
SMILES CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=CC(=C(C(=C3)O)C(=O)CCC4=CC=C(C=C4)O)O)CO)O)O)O)O)O

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in DMSO
Formula Weight 582.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Naringin dihydrochalcone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.