Learn More
Cayman Chemical NarlaprevIr 10mg
SDP
Catalog No. 502945249
Encompass_Preferred
An HCV NS3 inhibitor (Ki = 6 nM); also inhibits SARS-CoV-2 Mpro (IC50 = 16.11 µM); inhibits HCV replication in a cell-based replicon assay (EC90 = 40 nM) and SARS-CoV-2 replication in Vero cells (EC50 = 7.43 µM)
Catalog No. 50-294-5249 Supplier Cayman Chemical Supplier No. 4248710 MG
An HCV NS3 inhibitor (Ki = 6 nM); also inhibits SARS-CoV-2 Mpro (IC50 = 16.11 µM); inhibits HCV replication in a cell-based replicon assay (EC90 = 40 nM) and SARS-CoV-2 replication in Vero cells (EC50 = 7.43 µM)