Learn More
Cayman Chemical Narlaprevir, Purity: ≥95%, CAS: 865466-24-6, 5 mg
SDP
Catalog No. 502978640
Encompass_Preferred
An HCV NS3 inhibitor (Ki = 6 nM); also inhibits SARS-CoV-2 Mpro (IC50 = 16.11 µM); inhibits HCV replication in a cell-based replicon assay (EC90 = 40 nM) and SARS-CoV-2 replication in Vero cells (EC50 = 7.43 µM)
Catalog No. 50-297-8640 Supplier Cayman Chemical Supplier No. 424875 MG
An HCV NS3 inhibitor (Ki = 6 nM); also inhibits SARS-CoV-2 Mpro (IC50 = 16.11 µM); inhibits HCV replication in a cell-based replicon assay (EC90 = 40 nM) and SARS-CoV-2 replication in Vero cells (EC50 = 7.43 µM)