Learn More
Fine Science Tools Narrow Pattern Forceps, Stainless Steel, Straight, 18 cm
SDP
Catalog No. NC1717340
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 2.4 x 1.6mm/ Alloy / Material: Stainless Steel/ Length: 18cm
Catalog No. NC1717340 Supplier Fine Science Tools Supplier No. 1100218
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 2.4 x 1.6mm/ Alloy / Material: Stainless Steel/ Length: 18cm