Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Narrow Stem Pipettes 4.0 ml, 3.0 ml Bulb draw, Bulk package
SDP

Catalog No. 501899408
Encompass_Preferred

These pipets are ideal for Technique control such as loading gel, PCR work DNA sample removal, protein assays, lipid extractions, atomic absorption spectroscopy-reagent prep, reaction chamber, storage ampoule, portable aliquoting.
Bulb Draw: 3.0 ml
Drops/mL: 25
OAL: 8.5 cm
Stem diameter: 2.5 cm
Package: Bulk

Catalog No. 50-189-9408 Supplier Electron Microscopy Sciences Supplier No. 7096422
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These pipets are ideal for Technique control such as loading gel, PCR work DNA sample removal, protein assays, lipid extractions, atomic absorption spectroscopy-reagent prep, reaction chamber, storage ampoule, portable aliquoting.
Bulb Draw: 3.0 ml
Drops/mL: 25
OAL: 8.5 cm
Stem diameter: 2.5 cm
Package: Bulk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.