Learn More
MSC Vikan Tube Brush, 0.5 in Diameter, Medium Bristle, Polyester, for Flexible Handle
Catalog No. 502839360
Encompass
This tube brush is specifically designed for effective cleaning of pipes in demanding environments such as dairies, breweries, and the fishing industry. Featuring durable polyester bristles and robust stainless steel components, it is engineered for use with flexible handles to reach difficult areas.
  • Medium bristle stiffness for thorough cleaning.
  • Constructed with polyester (PBT) bristles and stainless steel (AISI 303Cu, AISI 304Cu) components.
  • Complies with EC 1935/2004 for food contact materials.
  • Meets EU Regulation 2023/2006/EC for Good Manufacturing Practice.
  • Complies with FDA Regulation CFR 21.
  • Meets UK 2019 No. 704 for food contact materials.
  • Compliant with REACH Regulation (EC) No. 1907/2006.
  • Complies with California Proposition 65.
  • Halal and Kosher compliant.
  • No intentionally added PFAS.
Catalog No. 50-283-9360 Supplier MSC Supplier No. 47173497
This tube brush is specifically designed for effective cleaning of pipes in demanding environments such as dairies, breweries, and the fishing industry. Featuring durable polyester bristles and robust stainless steel components, it is engineered for use with flexible handles to reach difficult areas.
  • Medium bristle stiffness for thorough cleaning.
  • Constructed with polyester (PBT) bristles and stainless steel (AISI 303Cu, AISI 304Cu) components.
  • Complies with EC 1935/2004 for food contact materials.
  • Meets EU Regulation 2023/2006/EC for Good Manufacturing Practice.
  • Complies with FDA Regulation CFR 21.
  • Meets UK 2019 No. 704 for food contact materials.
  • Compliant with REACH Regulation (EC) No. 1907/2006.
  • Complies with California Proposition 65.
  • Halal and Kosher compliant.
  • No intentionally added PFAS.