Learn More
MSC Vikan Tube Brush, 0.5000 in, 6-1/2 in Head, Polyester Bristles
Catalog No. 502835518
Encompass
This Vikan tube brush is designed for use with flexible handles, featuring stiff polyester bristles. It has a 0.5000 inch diameter and a 6-1/2 inch head length, making it suitable for various cleaning applications.
  • Stiff bristle flexibility for effective cleaning.
  • Durable polyester bristle material.
  • White bristle color.
  • 6-1/2 inch head length for extended reach.
  • 0.5000 inch diameter for narrow tubes.
Catalog No. 50-283-5518 Supplier MSC Supplier No. 47173505
This Vikan tube brush is designed for use with flexible handles, featuring stiff polyester bristles. It has a 0.5000 inch diameter and a 6-1/2 inch head length, making it suitable for various cleaning applications.
  • Stiff bristle flexibility for effective cleaning.
  • Durable polyester bristle material.
  • White bristle color.
  • 6-1/2 inch head length for extended reach.
  • 0.5000 inch diameter for narrow tubes.