Learn More
MSC Vikan Lobby Broom, Soft/Stiff, Polyester, 10.2", White
Catalog No. 502835858
Encompass
This lobby broom is engineered for effective cleaning in narrow spaces and dry areas, making it an ideal companion for a lobby pan. Constructed with durable polypropylene and soft/stiff polyester bristles, it offers robust performance and can be used with recommended handles.
  • Designed for cleaning in narrow spaces.
  • Ideal for sweeping dry areas and for use with a lobby pan.
  • Features soft/stiff polyester bristles for versatile cleaning.
  • Constructed from polypropylene, polyester, and stainless steel for durability.
  • Complies with EC 1935/2004 and FDA CFR 21 for food contact.
  • Autoclavable up to 249.8 °F and dishwasher safe up to 199.4 °F.
  • Suitable for use from -4 °F to 212 °F for non-food contact applications.
Catalog No. 50-283-5858 Supplier MSC Supplier No. 84116607
This lobby broom is engineered for effective cleaning in narrow spaces and dry areas, making it an ideal companion for a lobby pan. Constructed with durable polypropylene and soft/stiff polyester bristles, it offers robust performance and can be used with recommended handles.
  • Designed for cleaning in narrow spaces.
  • Ideal for sweeping dry areas and for use with a lobby pan.
  • Features soft/stiff polyester bristles for versatile cleaning.
  • Constructed from polypropylene, polyester, and stainless steel for durability.
  • Complies with EC 1935/2004 and FDA CFR 21 for food contact.
  • Autoclavable up to 249.8 °F and dishwasher safe up to 199.4 °F.
  • Suitable for use from -4 °F to 212 °F for non-food contact applications.