Learn More
Tritech Research Inc Narrow vial tray only (50/cs)
SDP
Catalog No. NC1086972
Encompass
Cardboard trays only (no dividers). 10 x 10 row configuration for narrow (25mm diameter) or shorty shell vials. 50 per case.
Catalog No. NC1086972 Supplier Tritech Research Inc Supplier No. T260T
Cardboard trays only (no dividers). 10 x 10 row configuration for narrow (25mm diameter) or shorty shell vials. 50 per case.