Learn More
Aviva Systems Biology Corp NARS Antibody - N-terminal region (ARP61484_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61484_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NARS. It was validated on Western Blot. . Alternate names: ASNRS, NARS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-394