Learn More
Cayman Chemical Narwedn 10mg
SDP
Catalog No. 502943936
Encompass_Preferred
An alkaloid with enzyme inhibitory activity; an inhibitor of AChE, BChE, and prolyl endopeptidase (IC50s = 281, 911, and 907 µM, respectively)
Catalog No. 50-294-3936 Supplier Cayman Chemical Supplier No. 4074410 MG
An alkaloid with enzyme inhibitory activity; an inhibitor of AChE, BChE, and prolyl endopeptidase (IC50s = 281, 911, and 907 µM, respectively)