Learn More
TELEFLEX LLC NASAL ATOMIZATION W/OUT SYRING
Catalog No. NC0725151
Encompass
LMA Mad nasal™ Intranasal Mucosal Atomization Device. MAD© Nasal Drug Delivery Device is a fast and effective way to deliver medications without needles. It gives you complete control of dosage, medication, and target as well as providing you with consistent results. Latex-free. This device has a Luer slip tip. Will work with any Luer syringe. 25 per box / 100 per case
Catalog No. NC0725151 Supplier TELEFLEX LLC Supplier No. MAD300
LMA Mad nasal™ Intranasal Mucosal Atomization Device. MAD© Nasal Drug Delivery Device is a fast and effective way to deliver medications without needles. It gives you complete control of dosage, medication, and target as well as providing you with consistent results. Latex-free. This device has a Luer slip tip. Will work with any Luer syringe. 25 per box / 100 per case