Learn More
Nasco Coliwasa Composite Liquid Waste Sampler

Description
- Unit can be used to sample most containerized liquid wastes, except those containing ketones, nitrobenzene, dimethylformamide, mesityl oxide and tetrahydrofuran
- Simple design is easy to use, and allows the rapid collection of samples, minimizing the exposure of the sample collector to potential hazards from the waste
- Manufactured of high-density polyethylene and designed to EPA standards
- Unit consists of a 1.53 m x 1.9 cm (5 ft. x 3/4 in.) ID tube and closing valve
- Sampler holds approximately 89mL per 0.31 m (3 oz. per ft.)
- Unit is not autoclavable
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 and ISO 13485:2003 |
| Volume (Metric) | 89 mL/0.31 m |
| For Use With (Application) | Used to sample most containerized liquid wastes |
| Includes | 5 ft. x 3/4 in. I.D. tube and closing valve |
| Material | HDPE |
| Type | Sludge Sampler |
| Length (English) | 5 ft. |
| Length (Metric) | 1.53 m |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.