Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whirl-Pak™ Meat and Poultry Carcass Sampling Kit (18 oz.) with PE Gloves and Buffer
GREENER_CHOICESDP

Catalog No. 018122 Shop All Whirl-Pak Products
Change view
Click to view available options
Capacity (Metric):
532 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01-812-2 532 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-812-2 Supplier Whirl-Pak™ Supplier No. B01324
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for sampling meat and turkey carcasses to test for the presence of E.coli.

  • One pair of non-handed (Size: L), sterile polyethylene gloves
  • 25 mL buffered peptone water (expiration date marked on the box)
  • Whirl-Pak™ Speci-Sponge™ bag
  • Two disposable sterile templates, one for meat carcasses (10 x 10 cm) and one for turkeys (5 x 10 cm)
  • Disposable template eliminates the need for re-sanitizing reusable templates, saving time and money and making it easier to transport the template to the carcass
  • Templates form to the contours of the carcass, making it easier to swab within the required area
  • Gloves and templates are packaged together in a sealed plastic bag that has a perforation for easy opening

Order Info

Box of 25 kits

Specifications

Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
For Use With (Application) Meat and Poultry Carcass Sampling
Includes One pair of non-handed (Size: L), sterile polyethylene gloves; 25 mL buffered peptone water (expiration date marked on the box); Whirl-Pak™ Speci-Sponge™ bag; Two disposable sterile templates, one for meat carcasses (10 x 10 cm) and one for turkeys (5 x 10 cm)
Material Polyethylene
Sterility Sterile
Fill Volume 18 oz.
Latex Free Yes
No. per Pack 25/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Plastic
Quantity 25/Pk.
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Length (English) 9 in.
Width (English) 4.25 in.
Product Type Sample Bag with Labeling Area
Capacity (English) 18 oz.
Capacity (Metric) 532 mL
Length (Metric) 23 cm
Width (Metric) 11.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.