Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whirl-Pak™ Write-On Plain Open Top Blender Bag, No Closure
GREENER_CHOICESDP

Catalog No. 01812231 Shop All Whirl-Pak Products
Change view
Click to view available options
Packaging Quantity:
250/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
01-812-231 250/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-812-231 Supplier Whirl-Pak™ Supplier No. B01474
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for quick testing and disposal.

  • Made of Whirl-Pak™ film
  • Each bag has a perimeter seal and is sealed at the top
  • Perforation for easy opening and to protect sterility, which allows for separate handling of each bag

Specifications

Product Type Sample Enrichment Bag
Capacity (Metric) 5441 mL
Dimensions (L x W) 50 x 37.5 cm (20 x 15 in.)
Blade Material Polypropylene
For Use With (Application) Quick Testing, Disposal
For Use With (Equipment) Stomacher, Blender, Homogenizer
Model B01474
Packaging Quantity 250/Box
Packaging Type Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.