Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Whirl-Pak™ Trans-Pak™ Non-Sterile Specimen Sample Bag
SDP

Catalog No. 01812124 Shop All Whirl-Pak Products
Change view
Click to view available options
Dimensions (L x W):
9 x 6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
01-812-124 9 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-812-124 Supplier Nasco Supplier No. B01253WA

missing translation for 'validMainframeMsg'

Unique specimen transport bag designed with an outside pouch to separate important paperwork from the inside of the bag - eliminating misplaced documents while protecting those handling the specimen

  • Outside pouch keeps important paperwork dry, clean, and separate from the sample.
  • Features the Biohazard symbol required by OSHA (Occupational Safety and Health Administration).

Specifications

Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Red
Disposable Single-use
For Use With (Application) Hazardous Waste Disposal
Includes Bag
Material Polyethylene
Sterility Sterile
Thickness (English) 2.50 mil
Dimensions (L x W) 9 x 6 in.
Length (English) 9 in.
Width (English) 6 in.
Product Type Biohazard Specimen Bag
Capacity (English) 24 oz.
Capacity (Metric) 710mL
Length (Metric) 23 cm
Width (Metric) 15 cm
Thickness (Metric) 0.064 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.