Learn More
MSC Nashville Wire Pallet Rack Welded Wire Decking, 42 x 46 in, 2700 lb Capacity
Catalog No. 5000536184
Encompass
This pallet rack welded wire decking is constructed from strong steel wire mesh, designed to integrate seamlessly with existing pallet rack systems. Its robust welded construction and gray baked enamel finish ensure durability and a professional appearance. It allows for efficient light, air, and sprinkler penetration, providing a clear view of upper tiers and minimizing dust and debris accumulation.
  • Constructed from strong 2-1/2 in. x 4 in. 6-gauge wire
  • Features welded construction with 13 gauge channel supports
  • Gray baked enamel finish provides durability
  • Permits light, air, and sprinkler penetration
  • Allows a clear view of upper tiers
  • Minimizes dust and debris accumulation
  • Installs easily by dropping into place over beams
  • Designed for use with beams that have 1-1/2 in. or 1-5/8 in. step
  • Offers a robust 2700 lb capacity
  • Made in USA
Catalog No. 50-005-36184 Supplier MSC Supplier No. 09496613
This pallet rack welded wire decking is constructed from strong steel wire mesh, designed to integrate seamlessly with existing pallet rack systems. Its robust welded construction and gray baked enamel finish ensure durability and a professional appearance. It allows for efficient light, air, and sprinkler penetration, providing a clear view of upper tiers and minimizing dust and debris accumulation.
  • Constructed from strong 2-1/2 in. x 4 in. 6-gauge wire
  • Features welded construction with 13 gauge channel supports
  • Gray baked enamel finish provides durability
  • Permits light, air, and sprinkler penetration
  • Allows a clear view of upper tiers
  • Minimizes dust and debris accumulation
  • Installs easily by dropping into place over beams
  • Designed for use with beams that have 1-1/2 in. or 1-5/8 in. step
  • Offers a robust 2700 lb capacity
  • Made in USA