Description
- Collect mucus and saliva samples then release them into the transport medium with a high recovery rate to detect bacterial and viral infections
- The US FDA and CDC recommend polyurethane foam swabs for PCR detection of most respiratory viruses including SARS-CoV-2 (COVID-19), Rhinoviruses and Enteroviruses (Picornaviridae), Influenza Viruses (Orthomyxoviridae), Parainfluenza, Metapneumoviruses and Respiratory Syncytial Viruses (Paramyxoviridae), Coronaviruses (Coronaviridae) and Bacterial Infections caused by Streptococcus Pneumoniae (Pneumonia) and Streptococcus Pyogenes (Strep Throat)
- Swabs utilize Foamtec developed polyurethane foam technology that provides an exceptional surface area to capture a maximum quantity of target fluids, while not bonding to those fluids to provide high subject recovery
- Cleaning processes and validated sterility ensure no background from RNAse and other contaminants for accurate test results
- Comfort in Nasopharyngeal swabbing is achieved with long, slender, and flexible handles keeping patient discomfort to a minimum
- Thin, high flexibility handles benefits the ability to reach remote areas of the nasal
septum and oral cavities - The plastic handle is engineered with a snap point to facilitate depositing the swab tip into a transport tube
- High surface area captures maximum amount of mucus and saliva
- No bonding to swabbed body fluids
- Validated sterile, clean swab surfaces
- High recovery rates of body fluids for accurate test results
- Maximum patient comfort
Specifications
Specifications
| Quantity | 1 Swab/Pk., 10 Pk./Box, 100 Boxes/Cs |
| Length (English) | 6.25 in. |
| For Use With (Application) | Collection of Mucus and Saliva Samples |
| Product Type | Nasopharyngeal (Nasal) Diagnostic Swab |
| Sterility | Sterile |
| Adhesive Type | Not Applicable |
| Handle Material | Plastic |
| Tip Material | Polyurethane Foam |