Learn More
Aviva Systems Biology Corp NASP Antibody - middle region (ARP57746_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57746_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NASP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NASP. Alternate names: DKFZp547F162, FLB7527, FLJ31599, FLJ35510, MGC19722, MGC20372, MGC2297, PRO1999ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62141