Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nuclear autoantigenic sperm protein (NASP) | 132989-56-1 | >95.0% | 10UG
SDP

Catalog No. 5000175122
Encompass_Preferred

Recombinant human nuclear autoantigenic sperm protein (NASP) expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for research use in biochemical and cell biology applications, including binding studies, structural analysis, and antibody production.

  • Produced in HEK293 mammalian expression system.
  • Contains a C-terminal His tag for affinity purification.
  • Available in small purified quantities (e.g., 5-100 μg).
  • Purity reported >95% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS (pH 7.4).
  • Suitable for biochemical assays, interaction studies, and antibody generation.

Catalog No. 50-001-75122 Supplier Medchemexpress LLC Supplier No. HYP70116310UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human nuclear autoantigenic sperm protein (NASP) expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for research use in biochemical and cell biology applications, including binding studies, structural analysis, and antibody production.

  • Produced in HEK293 mammalian expression system.
  • Contains a C-terminal His tag for affinity purification.
  • Available in small purified quantities (e.g., 5-100 μg).
  • Purity reported >95% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS (pH 7.4).
  • Suitable for biochemical assays, interaction studies, and antibody generation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.