Learn More
Medchemexpress LLC Nuclear autoantigenic sperm protein (NASP), human | >95.0% | 50UG
SDP
Catalog No. 5000176356
Encompass_Preferred
NASP Protein, Human (HEK293, His) is a recombinant human nuclear autoantigenic sperm protein produced in HEK293 mammalian cells and provided as a His-tagged, lyophilized preparation for research use. The mammalian expression system preserves native post-translational modifications, making the protein suitable for biochemical characterization, antibody production, and interaction studies.
  • Mammalian expression preserves native post-translational modifications.
  • His tag enables affinity purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Supplied lyophilized from PBS, pH 7.4 for stability and long-term storage.
  • Available in microgram-scale sizes for small-scale research applications.
  • Suitable for biochemical assays, binding studies, and antibody generation.
Catalog No. 50-001-76356 Supplier Medchemexpress LLC Supplier No. HYP70116350UG
NASP Protein, Human (HEK293, His) is a recombinant human nuclear autoantigenic sperm protein produced in HEK293 mammalian cells and provided as a His-tagged, lyophilized preparation for research use. The mammalian expression system preserves native post-translational modifications, making the protein suitable for biochemical characterization, antibody production, and interaction studies.
  • Mammalian expression preserves native post-translational modifications.
  • His tag enables affinity purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Supplied lyophilized from PBS, pH 7.4 for stability and long-term storage.
  • Available in microgram-scale sizes for small-scale research applications.
  • Suitable for biochemical assays, binding studies, and antibody generation.