Learn More
Medchemexpress LLC HY-12506A 5mg Medchemexpress, Naspm (trihydrochloride) CAS:1049731-36-3 Purity:>98%
SDP
Catalog No. 502081066
Encompass_Preferred
Medchemexpress, HY-12506A 5mg Naspm (trihydrochloride) CAS:1049731-36-3 Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1066 Supplier Medchemexpress LLC Supplier No. HY12506A5MG
Medchemexpress, HY-12506A 5mg Naspm (trihydrochloride) CAS:1049731-36-3 Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.