Learn More
Cayman Chemical NASPM (hydrochloride), Purity: ≥95%, CAS: 1049731-36-3, 10 mg
SDP
Catalog No. 502966046
Encompass_Preferred
A synthetic analog of Joro spider toxin that blocks GluR2-lacking, Ca2+-permeable AMPA receptors (IC50 = 0.33 µM) at -60 mV but not at +40 mV; protects against ischemia-induced neuronal cell death by preventing influx of toxic levels of Ca2+ into injured neurons
Catalog No. 50-296-6046 Supplier Cayman Chemical Supplier No. 1845310 MG
A synthetic analog of Joro spider toxin that blocks GluR2-lacking, Ca2+-permeable AMPA receptors (IC50 = 0.33 µM) at -60 mV but not at +40 mV; protects against ischemia-induced neuronal cell death by preventing influx of toxic levels of Ca2+ into injured neurons