Learn More
Cayman Chemical Nat-20(S)-yne, Purity: ≥90%, CAS: 1397692-46-4, 10 mg
SDP
Catalog No. 502894950
Encompass_Preferred
A form of nat-20(S)-OHC with a terminal alkyne group, which can be used in linking reactions known as click chemistry; has been conjugated with magnetic beads to demonstrate that nat-20(S)-OHC directly binds SMO
Catalog No. 50-289-4950 Supplier Cayman Chemical Supplier No. 900136910 MG
A form of nat-20(S)-OHC with a terminal alkyne group, which can be used in linking reactions known as click chemistry; has been conjugated with magnetic beads to demonstrate that nat-20(S)-OHC directly binds SMO