Learn More
Yamato Scientific America, Inc. Yamato 153L Natural Convection Oven, 115V
Catalog No. 501897979 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Standard 153L natural convection constant temperature drying oven with extensive features and simple operation. Economical and cost saving. Easy to use and maintain. Excellent temperature accuracy. Digital PID controller, calibration off-set and increased safety and self diagnostic functions. Operating temp ambient +5~280°C
Catalog No. 50-189-7979 Supplier Yamato Scientific America, Inc. Supplier No. DX602C
Standard 153L natural convection constant temperature drying oven with extensive features and simple operation. Economical and cost saving. Easy to use and maintain. Excellent temperature accuracy. Digital PID controller, calibration off-set and increased safety and self diagnostic functions. Operating temp ambient +5~280°C