Learn More
Yamato Scientific America, Inc. Yamato 92L Natural Convection Glassware Drying Oven, 220V
Catalog No. 501897987 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
92L glassware drying natural convection oven with large observation window for easy observation. Highly efficient heat insulation material for both internal and external structure. Adjustable foot for stability on uneven floors. Equipped with stainless steel pipe heater and water receiving plate at the bottom. Stainless steel interior, easy to clean and highly resistant to corrosion. Dial setting and digital display of temperature control and timer. Can be used to store instruments after drying. Operating temp ambient +5~70°C.
Catalog No. 50-189-7987 Supplier Yamato Scientific America, Inc. Supplier No. DG410C
92L glassware drying natural convection oven with large observation window for easy observation. Highly efficient heat insulation material for both internal and external structure. Adjustable foot for stability on uneven floors. Equipped with stainless steel pipe heater and water receiving plate at the bottom. Stainless steel interior, easy to clean and highly resistant to corrosion. Dial setting and digital display of temperature control and timer. Can be used to store instruments after drying. Operating temp ambient +5~70°C.