Learn More
AVANTOR MASTERFLEX (VWR LLC) Barbed fittings, Reducing Connector, Natural PP, 1/2"" x 1/4"" ID, 1/8"", 2-15/32""
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4061010/DEL
Encompass
Barbed fittings, Reducing Connector, Natural PP, 1/2" x 1/4" ID, 1/8", 2-15/32", Ideal for low-pressure applicationsQuick and easy assembly without toolsChemical resistantTemperture range 9 to 180¿F (-13 to 66¿C)Tubing ID 1/2" x 1/4"
Catalog No. NC1183822