Learn More
Pickering Laboratories Inc NAT SAMP AA STDBAS 0.1N HCL5ML
SDP
Catalog No. NC9054997
Encompass_Preferred
Native Sample Standard, Basics, in 0.1 N HCl, 2.5 µmole/mL, each (5 mL/bottle) Amino Acid Analysis.
Catalog No. NC9054997 Supplier Pickering Laboratories Inc Supplier No. 17000175
Native Sample Standard, Basics, in 0.1 N HCl, 2.5 µmole/mL, each (5 mL/bottle) Amino Acid Analysis.