Learn More
Medchemexpress LLC NAT1 Human His 50ug
SDP
Catalog No. 5000185048
Encompass_Preferred
Recombinant human N-acetyltransferase 1 (NAT1) with an N-terminal His tag, corresponding to UniProt accession P18440 (amino acids M1-I290). The product is supplied in small research sizes (for example, 50 μg), and is characterized by an apparent molecular weight of ~42 kDa and ≥95% purity, making it suitable for biochemical characterization and downstream applications.
  • High purity (≥95%) suitable for biochemical assays.
  • Contains His tag for affinity purification and detection.
  • Corresponds to full-length human NAT1 (M1-I290).
  • Apparent molecular weight ~42 kDa by SDS-PAGE.
  • Available in small research sizes for assay development.
  • Intended for research use in biochemical and structural studies.
Catalog No. 50-001-85048 Supplier Medchemexpress LLC Supplier No. HYP70542250UG
Recombinant human N-acetyltransferase 1 (NAT1) with an N-terminal His tag, corresponding to UniProt accession P18440 (amino acids M1-I290). The product is supplied in small research sizes (for example, 50 μg), and is characterized by an apparent molecular weight of ~42 kDa and ≥95% purity, making it suitable for biochemical characterization and downstream applications.
  • High purity (≥95%) suitable for biochemical assays.
  • Contains His tag for affinity purification and detection.
  • Corresponds to full-length human NAT1 (M1-I290).
  • Apparent molecular weight ~42 kDa by SDS-PAGE.
  • Available in small research sizes for assay development.
  • Intended for research use in biochemical and structural studies.