Learn More
Aviva Systems Biology Corp NAT13 Antibody - C-terminal region (ARP48934_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48934_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NAT13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NAT13. Alternate names: FLJ13194, MAK3, NAT5, SAN, hNAT5, hSAN, NAT13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53566