Learn More
Aviva Systems Biology Corp NAT15 Antibody - middle region (ARP48551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ14154. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NAT15. Alternate names: FLJ11693, NAT15ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53901