Learn More
Aviva Systems Biology Corp NAT2 Antibody - middle region (ARP44195_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44195_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NAT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NAT2. Alternate names: AAC2, PNATApplicationWB. Species reactivity: Human
Catalog No. 50-014-58743