Learn More
Aviva Systems Biology Corp NAT8L Antibody - C-terminal region (ARP49151_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49151_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NAT8L. It was validated on Western Blot. . Alternate names: CML3, FLJ37478, MGC117272, NAT8-LIKE, NACEDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-402