Learn More
Aviva Systems Biology Corp NAT9 Antibody - N-terminal region (ARP48780_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NAT9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NAT9. Alternate names: DKFZP564C103, EBSP, hNATLApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53900