Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Natalizumab | 189261-10-7 | ≥99.0% | 146.22 kDa | 25 MG
SDP

Catalog No. 5000347329
Encompass_Preferred

Natalizumab | 189261-10-7 | ≥99.0% | 146.22 kDa | 25 MG

Catalog No. 50-003-47329 Supplier Medchemexpress LLC Supplier No. 25MGHY108831A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Natalizumab | 189261-10-7 | ≥99.0% | 146.22 kDa | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.