Learn More
Selleck Chemical LLC Natamycin S1517-1g
SDP
Catalog No. 503110238
Encompass_Preferred
Natamycin (Pimaricin) a natural and versatile anti-fungal agent during fermentation by the bacterium Streptomyces natalensis commonly found in soil with little to no flavour interference
Catalog No. 50-311-0238 Supplier Selleck Chemical LLC Supplier No. S15171G
Natamycin (Pimaricin) a natural and versatile anti-fungal agent during fermentation by the bacterium Streptomyces natalensis commonly found in soil with little to no flavour interference