Learn More
Medchemexpress LLC Natamycin | 7681-93-8 | 99.1% | 200 MG
SDP
Catalog No. 5000450689
Encompass_Preferred
Natamycin is a macrolide antibiotic agent produced by several Streptomyces strains. It functions as a food preservative and an antifungal agent in agriculture, and is commonly used for fungal keratitis research. It inhibits the growth of fungi by preventing amino acid and glucose transport across the plasma membrane. Its mode of action does not rely on ergosterol-dependent permeabilization of the plasma membrane.
  • Inhibits fungal growth
  • Functions as a food preservative
  • Used as an antifungal agent in agriculture
  • Useful for fungal keratitis research
  • Does not rely on ergosterol-dependent permeabilization for its mode of action
Catalog No. 50-004-50689 Supplier Medchemexpress LLC Supplier No. HYB0133200MG
Natamycin is a macrolide antibiotic agent produced by several Streptomyces strains. It functions as a food preservative and an antifungal agent in agriculture, and is commonly used for fungal keratitis research. It inhibits the growth of fungi by preventing amino acid and glucose transport across the plasma membrane. Its mode of action does not rely on ergosterol-dependent permeabilization of the plasma membrane.
  • Inhibits fungal growth
  • Functions as a food preservative
  • Used as an antifungal agent in agriculture
  • Useful for fungal keratitis research
  • Does not rely on ergosterol-dependent permeabilization for its mode of action