Learn More
Medchemexpress LLC Natamycin | 7681-93-8 | 99.49% | 25 MG
SDP
Catalog No. 5000405805
Encompass_Preferred
Natamycin (Standard) is the analytical standard of Natamycin, intended for research and analytical applications. It is a macrolide antibiotic agent produced by several *Streptomyces* strains that inhibits the growth of fungi by inhibiting amino acid and glucose transport across the plasma membrane. It is used as a food preservative, an antifungal agent in agriculture, and for fungal keratitis research.
  • Used as a reference standard for assays
  • Used in qualitative, quantitative and methodological research experiments
  • Used in HPLC, GC and MS applications
  • Food preservative
  • Antifungal agent in agriculture
  • Used for fungal keratitis research
Catalog No. 50-004-05805 Supplier Medchemexpress LLC Supplier No. 25MGHYB0133R
Natamycin (Standard) is the analytical standard of Natamycin, intended for research and analytical applications. It is a macrolide antibiotic agent produced by several *Streptomyces* strains that inhibits the growth of fungi by inhibiting amino acid and glucose transport across the plasma membrane. It is used as a food preservative, an antifungal agent in agriculture, and for fungal keratitis research.
  • Used as a reference standard for assays
  • Used in qualitative, quantitative and methodological research experiments
  • Used in HPLC, GC and MS applications
  • Food preservative
  • Antifungal agent in agriculture
  • Used for fungal keratitis research