Learn More
Medchemexpress LLC Natamycin | 7681-93-8 | 99.5% | 100 MG
SDP
Catalog No. 5000450877
Encompass_Preferred
Natamycin (Pimaricin) is a macrolide antibiotic produced by Streptomyces strains. This analytical standard inhibits fungal growth by affecting amino acid and glucose transport across the plasma membrane. It is widely used as a food preservative, an antifungal agent in agriculture, and in fungal keratitis research. Intended for research and analytical applications, it supports qualitative, quantitative, and methodological experiments in techniques such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Inhibits growth of fungi
  • Used as a food preservative and antifungal agent
  • Supports fungal keratitis research
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-50877 Supplier Medchemexpress LLC Supplier No. HYB0133R100MG
Natamycin (Pimaricin) is a macrolide antibiotic produced by Streptomyces strains. This analytical standard inhibits fungal growth by affecting amino acid and glucose transport across the plasma membrane. It is widely used as a food preservative, an antifungal agent in agriculture, and in fungal keratitis research. Intended for research and analytical applications, it supports qualitative, quantitative, and methodological experiments in techniques such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Inhibits growth of fungi
  • Used as a food preservative and antifungal agent
  • Supports fungal keratitis research
  • Suitable for HPLC, GC, and MS experiments