Learn More
Medchemexpress LLC Natamycin | 7681-93-8 | MFCD00078278 | ≥98% | 5 G
SDP
Catalog No. 5000450942
Encompass_Preferred
Natamycin (Pimaricin) is a macrolide antibiotic agent produced by several *Streptomyces* strains. It inhibits the growth of fungi via inhibition of amino acid and glucose transport across the plasma membrane. It serves as a food preservative, an antifungal agent in agriculture, and is extensively used in fungal keratitis research.
  • Acts as a macrolide antibiotic agent.
  • Inhibits fungal growth by blocking amino acid and glucose transport.
  • Functions as a food preservative.
  • Used as an antifungal agent in agriculture.
  • Applicable in fungal keratitis research.
Catalog No. 50-004-50942 Supplier Medchemexpress LLC Supplier No. HYB01335G
Natamycin (Pimaricin) is a macrolide antibiotic agent produced by several *Streptomyces* strains. It inhibits the growth of fungi via inhibition of amino acid and glucose transport across the plasma membrane. It serves as a food preservative, an antifungal agent in agriculture, and is extensively used in fungal keratitis research.
  • Acts as a macrolide antibiotic agent.
  • Inhibits fungal growth by blocking amino acid and glucose transport.
  • Functions as a food preservative.
  • Used as an antifungal agent in agriculture.
  • Applicable in fungal keratitis research.