Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Natamycin (pimaricin) | 7681-93-8 | MFCD00135085 | 99.5% | 665.73 g/mol | C33H47NO13 | 10 MG
SDP

Catalog No. 5000298232
Encompass_Preferred

Natamycin (pimaricin) analytical standard provided as a 10 mg reference material for research and analytical applications. It is a macrolide antifungal agent commonly used as a reference in assays and method development.

  • Analytical standard suitable for research and QC applications.
  • High purity (99.49% reported; rounded to 99.5%).
  • Molecular weight 665.73 g/mol for accurate reference calculations.
  • CAS number 7681-93-8 for unambiguous identification.
  • Available in small milligram pack sizes for analytical workflows.

Catalog No. 50-002-98232 Supplier Medchemexpress LLC Supplier No. HYB0133R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Natamycin (pimaricin) analytical standard provided as a 10 mg reference material for research and analytical applications. It is a macrolide antifungal agent commonly used as a reference in assays and method development.

  • Analytical standard suitable for research and QC applications.
  • High purity (99.49% reported; rounded to 99.5%).
  • Molecular weight 665.73 g/mol for accurate reference calculations.
  • CAS number 7681-93-8 for unambiguous identification.
  • Available in small milligram pack sizes for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.