Learn More
Medchemexpress LLC Natamycin (standard) | 7681-93-8 | 99.5% | 50 MG
SDP
Catalog No. 5000450924
Encompass_Preferred
Natamycin (Standard) is an analytical standard, also known as Pimaricin. This macrolide antibiotic agent is produced by several Streptomyces strains. It inhibits fungal growth by preventing amino acid and glucose transport across the plasma membrane, making it suitable for various research and analytical applications.
  • Intended for research and analytical applications
  • Inhibits fungal growth by preventing amino acid and glucose transport
  • Used as a food preservative
  • Functions as an antifungal agent in agriculture
  • Utilized for fungal keratitis research
  • Applied in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Serves as a reference standard assay
Catalog No. 50-004-50924 Supplier Medchemexpress LLC Supplier No. HYB0133R50MG
Natamycin (Standard) is an analytical standard, also known as Pimaricin. This macrolide antibiotic agent is produced by several Streptomyces strains. It inhibits fungal growth by preventing amino acid and glucose transport across the plasma membrane, making it suitable for various research and analytical applications.
  • Intended for research and analytical applications
  • Inhibits fungal growth by preventing amino acid and glucose transport
  • Used as a food preservative
  • Functions as an antifungal agent in agriculture
  • Utilized for fungal keratitis research
  • Applied in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Serves as a reference standard assay