Learn More
Medchemexpress LLC Nateglinide | 105816-04-4 | MFCD00875706 | 99.95% | 100 MG
SDP
Catalog No. 5000408471
Encompass_Preferred
Nateglinide is a D-phenylalanine derivative that acts as an orally active, short-acting insulinotropic agent and a DPP IV inhibitor. It functions by inhibiting ATP-sensitive K+ channels in pancreatic β-cells and is utilized in the treatment of type 2 (non-insulin-dependent) diabetes mellitus. This product is intended for research use only and is not sold to patients.
  • Acts as an orally active, short-acting insulinotropic agent
  • Functions as a DPP IV inhibitor
  • Inhibits ATP-sensitive K+ channels in pancreatic β-cells
  • Used for the treatment of type 2 diabetes mellitus
  • Inhibits dinitrophenol-induced KATP currents
  • Stimulates human C-peptide secretion
  • Improves postprandial glucose concentrations
Catalog No. 50-004-08471 Supplier Medchemexpress LLC Supplier No. HYB0422100MG
Nateglinide is a D-phenylalanine derivative that acts as an orally active, short-acting insulinotropic agent and a DPP IV inhibitor. It functions by inhibiting ATP-sensitive K+ channels in pancreatic β-cells and is utilized in the treatment of type 2 (non-insulin-dependent) diabetes mellitus. This product is intended for research use only and is not sold to patients.
  • Acts as an orally active, short-acting insulinotropic agent
  • Functions as a DPP IV inhibitor
  • Inhibits ATP-sensitive K+ channels in pancreatic β-cells
  • Used for the treatment of type 2 diabetes mellitus
  • Inhibits dinitrophenol-induced KATP currents
  • Stimulates human C-peptide secretion
  • Improves postprandial glucose concentrations